Synonyms:
Heptacosafluorotributylamine Tris(nonafluorobutyl)amine Perfluorotributylamine FC-43
P0074-100G
P0074-25G
TCP0074-25GEA
84.76
USD
InStock
TCP0074-25G
TCP0074-100G
Tris(perfluorobutyl)amine 60.0% (by GC)
Tris(perfluorobutyl)amine
[for Mass spectrometry]
CAS Number: 311-89-7
MDL Number: MFCD00000436
Molecular Formula: C12F27N
Molecular Weight: 671.10
Purity/Analysis Method: 60.0% (GC)
Form: Clear Liquid
Boiling point (°C): 178
Specific Gravity (20/20): 1.89
Product Categories: Synthetic Organic Chemistry, Fluorous Chemistry, Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: Perfluorotributylamine, Tris(nonafluorobutyl)amine