Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tris(perfluorobutyl)amine 60.0% (by GC)

Supplier: TCI America
Synonyms: Heptacosafluorotributylamine Tris(nonafluorobutyl)amine Perfluorotributylamine FC-43

P0074-100G P0074-25G
TCP0074-25GEA 84.76 USD InStock
TCP0074-25G TCP0074-100G
Tris(perfluorobutyl)amine 60.0% (by GC)
Tris(perfluorobutyl)amine
[for Mass spectrometry]
CAS Number: 311-89-7
MDL Number: MFCD00000436
Molecular Formula: C12F27N
Molecular Weight: 671.10
Purity/Analysis Method: 60.0% (GC)
Form: Clear Liquid
Boiling point (°C): 178
Specific Gravity (20/20): 1.89

Product Categories: Synthetic Organic Chemistry, Fluorous Chemistry, Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: Perfluorotributylamine, Tris(nonafluorobutyl)amine


Formula: C₁₂F₂₇N
MW: 671.1 g/mol
Boiling Pt: 178 °C
Density: 1.89 (20 °C)
MDL Number: MFCD00000436
CAS Number: 311-89-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/07/75/28990775.jpg
Order Now
Specification Test Results
Purity/Analysis Method 60.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty