Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphorus pentoxide - methanesulfonic acid

Supplier: TCI America
Synonyms: Eaton's Reagent

P1042-25G P1042-500G
TCP1042-500GEA 182.97 USD InStock
TCP1042-500G TCP1042-25G
Phosphorus pentoxide - methanesulfonic acid
Phosphorus pentoxide - methanesulfonic acid
CAS Number: 39394-84-8
MDL Number: MFCD00070545
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 122
Flash Point (°C): 113

Other Product Names / Synonyms: Eaton's Reagent


MW: 141.95 g/mol
Boiling Pt: 122 °C
Flash Pt: 113 °C
MDL Number: MFCD00070545
CAS Number: 39394-84-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/16/79/28991679.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty