Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Phenyl-3H-1,2,4-triazole-3,5-(4H)-dione

Supplier: TCI America
Synonyms: PTAD4-Phenyl-1,2,4-triazoline-3,5-dione4-Phenyl-3H-1,2,4-triazole-3,5(4H)-dione3,5-Dihydro-4-phenyl-4H-1,2,4-triazole-3,5-dione

P1184-1G P1184-5G
TCP1184-001GEA 129.02 USD InStock
TCP1184-001G TCP1184-5G
4-Phenyl-3H-1,2,4-triazole-3,5-(4H)-dione
4-Phenyl-3H-1,2,4-triazole-3,5(4H)-dione
CAS Number: 4233-33-4
MDL Number: MFCD00003148
Molecular Formula: C8H5N3O2
Molecular Weight: 175.15
Form: Crystal
Color: Red
Storage Temperature: 0°C

Other Product Names / Synonyms: PTAD


Formula: C₈H₅N₃O₂
MW: 175.1400 g/mol
MDL Number: MFCD00003148
CAS Number: 4233-33-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/18/15/28991815.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty