Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Palladium(II) bromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: Palladium dibromide

P1937-2G
TCP1937-2GEA 605 USD InStock
TCP1937-2G
Palladium(II) bromide ≥98.0% (by titrimetric analysis)
Palladium (II) bromide
CAS Number: 13444-94-5
MDL Number: MFCD00011170
Molecular Formula: PdBr2
Molecular Weight: 266.23
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: Gray

Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts
Other Product Names / Synonyms: Palladium Dibromide


Formula: Br₂Pd
MW: 266.23 g/mol
MDL Number: MFCD00011170
CAS Number: 13444-94-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/31/97/28993197.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty