Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycerine ≥99.0%

Supplier: TCI America
Synonyms: GlysaninGlycerinGlyceritolPropane-1,2,3-triolOsmoglynPropanetriolGlycerolGlyrolGlycyl alcohol1,2,3-PropanetriolTrihydroxypropane

S0373-10G S0373-1G
TCS0373-1GEA 27.41 USD InStock
TCS0373-1G TCS0373-10G
Glycerine ≥99.0%
Glycerine
[Matrix for FABMS and liquid SIMS]
CAS Number: 56-81-5
MDL Number: MFCD00004722
Molecular Formula: C3H8O3
Molecular Weight: 92.09
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 290
Flash Point (°C): 177
Specific Gravity (20/20): 1.26

Product Categories: Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: Glycerin


Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290 °C
Density: 1.26 (20 °C)
Flash Pt: 177 °C
MDL Number: MFCD00004722
CAS Number: 56-81-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/40/88/28994088.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty