Would you like to log in again?
Magic Bullet [mixture of Dithiothreitol and Dithioerythritol (3:1)]
Supplier: TCI America
S0375-1G
TCS0375-001GEA 135 USD InStock
TCS0375-001G
Magic Bullet [mixture of Dithiothreitol and Dithioerythritol (3:1)]
Magic bullet [mixture of dithiothreitol and dithioerythritol (3:1)]
[Matrix for FABMS and liquid SIMS]
MDL Number: MFCD00147515
Product Categories: Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty