Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Nitrophenyl octyl ether ≥98.0%

Supplier: TCI America
Synonyms: 1-Nitro-2-(n-octyloxy)benzene2-(n-Octyloxy)nitrobenzene1-Nitro-2-(octyloxy)benzene

S0380-1G S0380-10G
TCS0380-1GEA 32.48 USD InStock
TCS0380-1G TCS0380-10G
2-Nitrophenyl octyl ether ≥98.0%
2-Nitrophenyl octyl ether
[Matrix for FABMS and liquid SIMS]
CAS Number: 37682-29-4
MDL Number: MFCD00014693
Molecular Formula: C14H21NO3
Molecular Weight: 251.33
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 198
Flash Point (°C): 113
Specific Gravity (20/20): 1.04


Formula: C₁₄H₂₁NO₃
MW: 251.32 g/mol
Boiling Pt: 198 °C
Density: 1.04 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00014693
CAS Number: 37682-29-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/40/92/28994092.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty