Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium azide ≥99.0% (by titrimetric analysis)

Supplier: TCI America

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Synonyms: Sodium triazide

S0489-100G
TCS0489-100GEA 63.08 USD InStock
TCS0489-100G
Sodium azide ≥99.0% (by titrimetric analysis)
Sodium azide
CAS Number: 26628-22-8
MDL Number: MFCD00003536
Molecular Formula: N3Na
Molecular Weight: 65.01
Purity/Analysis Method: 99.0% (T)
Form: Crystal
Color: White


Formula: NaN₃
MW: 65.01 g/mol
MDL Number: MFCD00003536
CAS Number: 26628-22-8
UN: 1687
ADR: 6.1,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/41/48/28994148.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (T)

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty