Synonyms:
4-[3,5-Dimethyl-4-(4-nitrobenzyloxy)phenyl]-4-oxobutyric Acid Succinimidyl Ester
S0808-100MG
TCS0808-100MGEA
233
USD
InStock
TCS0808-100MG
Succinimidyl-4-[3,5-dimethyl-4-(4-nitrobenzyloxy)phenyl]-4-oxobutyrate ≥96.0% (by HPLC)
Succinimidyl-4-[3,5-dimethyl-4-(4-nitrobenzyloxy)phenyl]-4-oxobutyrate
CAS Number: 948995-62-8
Molecular Formula: C23H22N2O8
Molecular Weight: 454.44
Purity/Analysis Method: 96.0% (HPLC)
Form: Crystal
Color: White
Storage Temperature: 0-10°C
Product Categories: Analytical Science, Derivatization Reagents for MS
Other Product Names / Synonyms: 4-[3,5-Dimethyl-4-(4-nitrobenzyloxy)phenyl]-4-oxobutyric Acid Succinimidyl Ester