Synonyms:
2-(4-Methylcyclohex-3-en-1-yl)propan-2-ol
, Terpineol
T0022-500ML
T0022-25ML
TCT0022-500MLEA
122.82
USD
InStock
TCT0022-500ML
TCT0022-025ML
(±)-α-Terpineol ≥80.0%
(±)-α-Terpineol
CAS Number: 98-55-5
MDL Number: MFCD00001557
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method: 80.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 101
Specific Gravity (20/20): 0.94
Product Categories: Bioscience, Monocyclic Monoterpenes
Other Product Names / Synonyms: 1-p-Menthen-8-ol