Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eosin Y (yellowish) ≥90.0% (by UV method)

Supplier: TCI America
Warning
Synonyms: Tetrabromofluorescein disodium salt Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate Eosin yellowish Eosin Y disodium salt 2,4,5,7-Tetrabromofluorescein disodium salt Solvent Red 43 Eosin 225 Field's Stain B

T0037-25G
TCT0037-025GEA 44.76 USD InStock
TCT0037-025G
Eosin Y (yellowish) ≥90.0% (by UV method)
Eosin Y (yellowish)
CAS Number: 17372-87-1
MDL Number: MFCD00005040
Molecular Formula: C20H8Br4O5
Molecular Weight: 691.86
Purity/Analysis Method: 90.0% (E)
Form: Crystal
Color: Yellow Red
Lambda max.: 517 nm (0.02mol/L AcONH4 sol.)

Product Categories: Materials Chemistry, Xanthene Dyes (Functional Organic Dyes), Bioscience, Stains & Dyes, Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: Eosine, Tetrabromofluorescein Sodium Salt


Formula: C₂₀H₆Br₄Na₂O₅
MW: 691.86 g/mol
Melting Pt: 295…296 °C
Storage Temperature: Ambient
MDL Number: MFCD00133309
CAS Number: 17372-87-1
EINECS: 241-409-6
Merck Index: 13,03634
https://us-legacy.vwr.com/stibo/web/std.lang.all/43/81/28994381.jpg
Order Now
Specification Test Results
Purity/Analysis Method 90.0% (E)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty