Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning
Synonyms: N,N,N',N'-Tetramethyl-1,4-phenylenediamine dihydrochlorideN1,N1,N4,N4-Tetramethylbenzene-1,4-diamine dihydrochlorideTMPD 2 HClN,N,N',N'-Tetramethyl-1,4-phenylenediammonium dichloride

T0151-5G T0151-1G T0151-25G
TCT0151-5GEA 47.71 USD InStock
TCT0151-5G TCT0151-001G TCT0151-025G
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride ≥98.0% (by HPLC, total nitrogen)
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride
CAS Number: 637-01-4
MDL Number: MFCD00012482
Molecular Formula: C10H16N2
Molecular Weight: 237.17
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 224

Other Product Names / Synonyms: TMPD


Formula: C₁₀H₁₈Cl₂N₂
MW: 237.17 g/mol
Melting Pt: 224 °C
Storage Temperature: Ambient
MDL Number: MFCD00012482
CAS Number: 637-01-4
EINECS: 211-274-8
Merck Index: 12,09367
https://us-legacy.vwr.com/stibo/web/std.lang.all/44/75/28994475.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,N)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty