Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetramethylsilane ≥99.0%

Supplier: TCI America
Danger
Synonyms: TMS

T0154-25ML T0154-500ML T0154-100ML
TCT0154-100MLEA 119.74 USD InStock
TCT0154-100ML TCT0154-25ML TCT0154-500ML
Tetramethylsilane ≥99.0%
Tetramethylsilane
[for NMR]
CAS Number: 75-76-3
MDL Number: MFCD00008274
Molecular Formula: C4H12Si
Molecular Weight: 88.23
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 26
Flash Point (°C): -27
Specific Gravity (20/20): 0.64
Storage Temperature: 0-10°C


Formula: C₄H₁₂Si
MW: 88.22 g/mol
Boiling Pt: 26 °C
Melting Pt: –99 °C
Density: 0.64 (20 °C)
Flash Pt: –27 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008274
CAS Number: 75-76-3
EINECS: 200-899-1
UN: 2749
ADR: 3,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/44/77/28994477.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty