Synonyms:
Chlorbutanol hemihydrate
, Chlorobutanol hydrous
, 1,1,1-Trichloro-2-methyl-2-propanol Hemihydrate
T0386-25G
T0386-500G
TCT0386-025GEA
32.46
USD
InStock
TCT0386-025G
TCT0386-500G
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate ≥98.0% (by GC, titration analysis)
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate
CAS Number: 6001-64-5
MDL Number: MFCD00004461
Molecular Formula: C4H7Cl3O
Molecular Weight: 177.45
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 175
Melting point (°C): 78
Flash Point (°C): 100
Other Product Names / Synonyms: Acetonechloroform, beta,beta,beta-Trichloro-tert-butyl Alcohol, Chloretone