Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,1,1-Trichloro-2-methyl-2-propanol hemihydrate ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Synonyms: Chlorbutanol hemihydrateChlorobutanol hydrous1,1,1-Trichloro-2-methyl-2-propanol Hemihydrate

T0386-25G T0386-500G
TCT0386-025GEA 32.46 USD InStock
TCT0386-025G TCT0386-500G
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate ≥98.0% (by GC, titration analysis)
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate
CAS Number: 6001-64-5
MDL Number: MFCD00004461
Molecular Formula: C4H7Cl3O
Molecular Weight: 177.45
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 175
Melting point (°C): 78
Flash Point (°C): 100

Other Product Names / Synonyms: Acetonechloroform, beta,beta,beta-Trichloro-tert-butyl Alcohol, Chloretone


Formula: C₈H₁₆Cl₆O₃
MW: 372,93 g/mol
Boiling Pt: 175 °C
Melting Pt: 78 °C
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD02179352
CAS Number: 6001-64-5
EINECS: 200-317-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/46/53/28994653.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty