Synonyms:
2-(4-Methylcyclohex-3-en-1-yl)propan-2-ol
, Terpineol
T0984-100ML
T0984-500ML
T0984-25ML
TCT0984-500MLEA
289.28
USD
InStock
TCT0984-500ML
TCT0984-025ML
TCT0984-100ML
(±)-α-Terpineol ≥95.0%
(±)-α-Terpineol
CAS Number: 98-55-5
MDL Number: MFCD00001557
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 218
Flash Point (°C): 101
Freezing point (°C): 31
Product Categories: Bioscience, Monocyclic Monoterpenes
Other Product Names / Synonyms: 1-p-Menthen-8-ol