Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4,5-Trimethoxytoluene ≥98.0%

Supplier: TCI America
Synonyms: 1,2,3-Trimethoxy-5-methylbenzene

T1236-500G T1236-25G
TCT1236-500GEA 265.93 USD InStock
TCT1236-500G TCT1236-025G
3,4,5-Trimethoxytoluene ≥98.0%
3,4,5-Trimethoxytoluene
CAS Number: 6443-69-2
MDL Number: MFCD00008397
Molecular Formula: C10H14O3
Molecular Weight: 182.22
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Flash Point (°C): 131
Freezing point (°C): 36


Formula: C₁₀H₁₄O₃
MW: 182.22 g/mol
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 35…37 °C
Density: 1.08 g/cm³ (20 °C)
Flash Pt: 131 °C
MDL Number: MFCD00008397
CAS Number: 6443-69-2
EINECS: 229-239-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/52/77/28995277.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty