Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')praseodymium(III)

Supplier: TCI America
Synonyms: Pr(TMHD)3Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)praseodymium(III)

T1264-1G
TCT1264-001GEA 145.92 USD InStock
TCT1264-001G
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')praseodymium(III)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')praseodymium(III)
[NMR Shift Reagent]
CAS Number: 15492-48-5
MDL Number: MFCD00000027
Molecular Formula: C33H57O6Pr
Molecular Weight: 690.72
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 220

Other Product Names / Synonyms: Praseodymium(III) Tris(2,2,6,6-tetramethyl-3,5-heptanedionate), Pr(dpm)3


Formula: C₃₃H₅₇O₆Pr
MW: 690.71 g/mol
Melting Pt: 220 °C
MDL Number: MFCD00000027
CAS Number: 15492-48-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/53/04/28995304.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty