Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')praseodymium(III) ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)praseodymium(III)Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)praseodymium(III)Pr(FOD)3

T1511-1G
TCT1511-001GEA 153.2 USD InStock
TCT1511-001G
Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')praseodymium(III) ≥97.0% (by titrimetric analysis)
Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')praseodymium(III)
[NMR Shift Reagent]
CAS Number: 17978-77-7
MDL Number: MFCD00064656
Molecular Formula: C30H30F21O6Pr
Molecular Weight: 1026.44
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 219

Product Categories: Analytical Science, Shift Reagents
Other Product Names / Synonyms: Praseodymium(III) Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)


Formula: C₃₀H₃₀F₂₁O₆Pr
MW: 1026.43 g/mol
Melting Pt: 219 °C
MDL Number: MFCD00064656
CAS Number: 17978-77-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/55/27/28995527.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty