Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4,6-Tri-tert-butylnitrosobenzene ≥98.0%

Supplier: TCI America

T1534-100MG T1534-1G
TCT1534-1GEA 710.29 USD InStock
TCT1534-1G TCT1534-100MG
2,4,6-Tri-tert-butylnitrosobenzene ≥98.0%
2,4,6-Tri-tert-butylnitrosobenzene
CAS Number: 24973-59-9
MDL Number: MFCD00008817
Molecular Formula: C18H29NO
Molecular Weight: 275.44
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: Pale Green
Melting point (°C): 168
Storage Temperature: 0-10°C


Formula: C₁₈H₂₉NO
Melting Pt: 168 °C
MDL Number: MFCD00008817
CAS Number: 24973-59-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/55/46/28995546.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty