Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,2,6,6-Tetramethylpiperidinooxy (TEMPO) ≥98.0% (by GC, titration analysis) free radical

Supplier: TCI America
Synonyms: 2,2,6,6-Tetramethylpiperidine 1-oxyl2,2,6,6-Tetramethyl-1-oxylpiperidineTEMPO

T1560-25G T1560-5G
TCT1560-25GEA 131.95 USD InStock
TCT1560-25G TCT1560-005G
2,2,6,6-Tetramethylpiperidinooxy (TEMPO) ≥98.0% (by GC, titration analysis) free radical
2,2,6,6-Tetramethylpiperidinooxy (TEMPO, free radical)
CAS Number: 2564-83-2
MDL Number: MFCD00009599
Molecular Formula: C9H18NO
Molecular Weight: 156.25
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Color: Reddish Yellow
Melting point (°C): 39
Flash Point (°C): 67
Storage Temperature: 0-10°C

Product Categories: Synthetic Organic Chemistry, C-H Bond Activation Reaction, Oxidation, Oxidation, Materials Chemistry, Organic Radicals (Magnetic Materials), Precise Polymerization Reagents, Analytical Science, Spin Labels
Other Product Names / Synonyms: TEMPO


Formula: C₉H₁₈NO
MW: 156.25 g/mol
Melting Pt: 39 °C
Flash Pt: 67 °C
MDL Number: MFCD00009599
CAS Number: 2564-83-2
EINECS: 219-888-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/55/71/28995571.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty