Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium-3-(trimethylsilyl)-1-propanesulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: 3-(Trimethylsilyl)-1-propanesulfonic Acid Sodium SaltDSSSodium 2,2-Dimethyl-2-silapentane-5-sulfonate

T1638-1G T1638-5G
TCT1638-1GEA 256.38 USD InStock
TCT1638-1G TCT1638-005G
Sodium-3-(trimethylsilyl)-1-propanesulfonate ≥98.0% (by titrimetric analysis)
Sodium-3-(trimethylsilyl)-1-propanesulfonate
[1H NMR Standard for D2O Solvent]
CAS Number: 2039-96-5
MDL Number: MFCD00007537
Molecular Formula: C6H16O3SSi
Molecular Weight: 218.32
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White

Product Categories: Analytical Science, Standards for NMR
Other Product Names / Synonyms: 3-(Trimethylsilyl)-1-propanesulfonic Acid Sodium Salt, DSS, Sodium 2,2-Dimethyl-2-silapentane-5-sulfonate


Formula: C₆H₁₆O₃SSi
MW: 218.32 g/mol
MDL Number: MFCD00007537
CAS Number: 2039-96-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/56/30/28995630.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty