Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5,6,8-Tetrabromo-1,10-phenanthroline ≥95.0% (by HPLC)

Supplier: TCI America

T3133-100MG
TCT3133-100MGEA 164 USD InStock
TCT3133-100MG
3,5,6,8-Tetrabromo-1,10-phenanthroline ≥95.0% (by HPLC)
3,5,6,8-Tetrabromo-1,10-phenanthroline
CAS Number: 66127-00-2
MDL Number: MFCD09879848
Molecular Formula: C12H4Br4N2
Molecular Weight: 495.79
Purity/Analysis Method: 95.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 356


Formula: C₁₂H₄Br₄N₂
MW: 495.79 g/mol
Melting Pt: 356 °C
MDL Number: MFCD09879848
CAS Number: 66127-00-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/67/30/28996730.jpg
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (HPLC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty