Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Deuterium oxide (99.8% D)

Supplier: TCI America
Synonyms: Heavy waterWater-D2

W0002-10ML W0002-100ML
TCW0002-10MLEA 68.05 USD InStock
TCW0002-10ML TCW0002-100ML
Deuterium oxide (99.8% D)
Deuterium oxide
CAS Number: 7789-20-0
MDL Number: MFCD00044636
Molecular Formula: D2O
Molecular Weight: 20.03
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 101
Melting point (°C): 4
Specific Gravity (20/20): 1.11

Other Product Names / Synonyms: Water-d2


Formula: D₂O
MW: 20 g/mol
Boiling Pt: 101 °C
Melting Pt: 4 °C
Density: 1.11 (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00044636
CAS Number: 7789-20-0
EINECS: 232-148-9
Merck Index: 12,02984
https://us-legacy.vwr.com/stibo/web/std.lang.all/73/58/28997358.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty