Synonyms:
Zirconocene dichloride
Z0007-5G
Z0007-25G
TCZ0007-5GEA
46.89
USD
InStock
TCZ0007-5G
TCZ0007-25G
Bis(cyclopentadienyl)zirconium dichloride ≥97.0% (by titrimetric analysis)
Bis(cyclopentadienyl)zirconium dichloride
CAS Number: 1291-32-3
MDL Number: MFCD00003726
Molecular Formula: C10H10Cl2Zr
Molecular Weight: 292.31
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Color: White
Product Categories: Synthetic Organic Chemistry, Glycosylation, Materials Chemistry, Polymerization Catalysts, Analytical Science, Chemical Shift Standards for Multinuclear NMR
Other Product Names / Synonyms: Bis(cyclopentadienyl)zirconium Dichloride