Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(cyclopentadienyl)zirconium dichloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: Zirconocene dichloride

Z0007-5G Z0007-25G
TCZ0007-5GEA 46.89 USD InStock
TCZ0007-5G TCZ0007-25G
Bis(cyclopentadienyl)zirconium dichloride ≥97.0% (by titrimetric analysis)
Bis(cyclopentadienyl)zirconium dichloride
CAS Number: 1291-32-3
MDL Number: MFCD00003726
Molecular Formula: C10H10Cl2Zr
Molecular Weight: 292.31
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Color: White

Product Categories: Synthetic Organic Chemistry, Glycosylation, Materials Chemistry, Polymerization Catalysts, Analytical Science, Chemical Shift Standards for Multinuclear NMR
Other Product Names / Synonyms: Bis(cyclopentadienyl)zirconium Dichloride


Formula: C₁₀H₁₀Cl₂Zr
MW: 292.32 g/mol
MDL Number: MFCD00003726
CAS Number: 1291-32-3
EINECS: 215-066-8
UN: 3261
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/30/28997430.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty