Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycylglycine (Gly-Gly)

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

12014-1000 12014-0250 12014-0010
TS12014-0250EA 53 USD InStock
TS12014-0250 TS12014-1000 TS12014-0010
Glycylglycine (Gly-Gly)
Proteins and Peptides
Glycylglycine (Gly-Gly) contains ≤0.1% sulphated ash, ≤100 ppm chloride and ≤5 ppm heavy metals (as Pb).

  • Infrared spectrum: Conforms
  • Loss on drying: ≤0.5% (105 °C, 4 h)
  • Titration with HClO₄: 99.0 to 101.0% (on dried basis)
  • EINECS: 209-127-8
  • Melting point: 262 to 264 °C (dec.)

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty