Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide standard solution 0.1 mol/l (0.1 N) in water

Supplier: Thermo Scientific Chemicals
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

12419-0025 124190250 124190010
TS124190250EA 214 USD InStock
TS124190250 TS124190010 TS12419-0025
Sodium hydroxide standard solution 0.1 mol/l (0.1 N) in water
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Density: 1 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
Order Now
Specification Test Results
Appearance Clear colorless Solution
Strength 0.0995 to 0.1005 N (20 °C)
Melting point 50 to 54 °C
GC ≥98.5 %
NMR (1H-NMR) Consistent with structure
Free alkali (KOH) ≤1 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty