Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid standard solution 1 mol/l (1 N) in water

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Muriatic acid

12421-0010
TS12421-0010EA 75.64 USD InStock
TS12421-0010
Hydrochloric acid standard solution 1 mol/l (1 N) in water
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Density: 1.02 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear colorless Solution
Strength 0.998 to 1.002 N (20°C)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty