Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lactic acid 90%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Hydroxypropionic acid2-Hydroxypropanoic acidDL-Lactic acid

12506-0100 12506-5000 12506-0010 12506-0025 12506-0250
TS12506-5000EA 84.56 USD InStock
TS12506-5000 TS12506-0250 TS12506-0025 TS12506-0010 TS12506-0100
Lactic acid 90%
Lactic acid


Formula: H₃CCH(OH)COOH
MW: 90.08 g/mol
Boiling Pt: 122 °C (15 mmHg)
Melting Pt: 18 °C
Density: 1.2 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
EINECS: 200-018-0
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4252 to 1.4272 (20°C, 589 nm)
Residue after ignition ≤0.1 %
Specific optical rotation -0.05° to +0.05° (25°C, 589 nm) (neat)
Acids (Citric,oxalic,phosphoric or tartaric) Passes test
Titration with NaOH 88.0 to 92.0 %
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm
Sugars Passes test

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty