Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 98%, pellets, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

134070250 13407-0010 13407-0100 134070025
TS13407-0100EA 239 USD InStock
TS13407-0100 TS13407-0010
Sodium hydroxide 98%, pellets, Extra Pure
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C (760 mmHg)
Melting Pt: 318 °C
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Order Now
Specification Test Results
Appearance White Pellets
Titration with HCl ≥97.5 %
Chloride (Cl) ≤50 ppm
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤40 ppm
Sodium carbonate (Na2CO3) ≤1.0 %
Aluminium (Al) ≤10 ppm
Calcium (Ca) ≤10 ppm
Iron (Fe) ≤5 ppm
Lead (Pb) ≤1 ppm
Nickel (Ni) ≤5 ppm
Potassium (K) ≤250 ppm
Silver (Ag) ≤0.5 ppm
Silicon dioxide (SiO2) ≤5 ppm
Nitrogen compounds (as N) ≤0.0005 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty