Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl acetate 99+%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: EtOAcAcetidinEthyl ethanoateVinegar naphthaEthyl acetic esterAcetic acid ethyl esterAcetic etherAcetoxyethane

14947-0250 14947-0050 14947-0025
TS14947-0025EA 110 USD InStock
TS14947-0025 TS14947-0250 TS14947-0050
Ethyl acetate 99+%, Extra Pure
Ethyl acetate


Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77.1 °C (1013 hPa)
Melting Pt: –83.5 °C
Density: 0.902 g/cm³
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
https://us-legacy.vwr.com/stibo/web/std.lang.all/20/72/722072.jpg
Order Now
Specification Test Results
Appearance Clear liquid
Water ≤0.05 % (K.F.)
GC ≥99.0 %
Color scale ≤10 APHA
Residue after evaporation ≤0.005 %
Residual solvents ≤10 % MTBE

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty