Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl ethyl ketone 99+%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-ButanoneMEKButan-2-oneEK 500 AdditiveButanoneEthyl methyl ketone

14967-0025 14967-0250
TS14967-0250EA 33 USD InStock
TS14967-0250 TS14967-0025
Methyl ethyl ketone 99+%, Extra Pure
Methyl ethyl ketone


Formula: C₂H₅COCH₃
MW: 72.11 g/mol
Boiling Pt: 80 °C (760 mmHg)
Melting Pt: –87 °C
Density: 0.806 g/cm³
Flash Pt: –7 °C
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3,II
REACH: 01-2119457290-43
Merck Index: 12,06149
https://us-legacy.vwr.com/stibo/web/std.lang.all/03/79/720379.jpg
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3780 to 1.3800 (20 °C, 589 nm)
Water ≤0.2 %
GC ≥99.0 %
Residue after evaporation ≤0.005 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty