Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eosin Y (yellowish), high purity biological stain

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Tetrabromofluorescein disodium saltEosin Y disodium saltEosin 2252′,4′,5′,7′-Tetrabromofluorescein disodium saltSolvent Red 43Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoateDisodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)Field's Stain B2,4,5,7-Tetrabromofluorescein disodium salt

15288-0250 15288-1000 15288-5000
TS15288-0250EA 46.75 USD InStock
TS15288-0250 TS15288-5000 TS15288-1000
Eosin Y (yellowish), high purity biological stain
Eosin Y (yellowish)


Formula: C₂₀H₆Br₄Na₂O₅
MW: 691.86 g/mol
Melting Pt: >300 °C (dec.)
Density: 1.018 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00005040
CAS Number: 17372-87-1
EINECS: 241-409-6
Order Now
Specification Test Results
Appearance Red to brown Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤10 %
Dye content ≥90 % (on dry substance)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty