Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diphenylboric acid-2-aminoethyl ester (2-APB) 96%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Aminoethyl diphenylborinate2-((Diphenylboryl)oxy)ethanamine

15542-0100 15542-0250 15542-0050
TS15542-0100EA 122 USD InStock
TS15542-0100 TS15542-0250 TS15542-0050
Diphenylboric acid-2-aminoethyl ester (2-APB) 96%
Diphenylboric acid 2-aminoethyl ester (2-APB)


Formula: C₁₄H₁₆BNO
MW: 225.1 g/mol
Boiling Pt: 150…154 °C (760 mmHg)
Melting Pt: 81…80 °C
Density: 0.84 g/cm³
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00014823
CAS Number: 524-95-8
EINECS: 203-857-0
UN: 3071
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4480 to 1.4520 (20°C, 589 nm)
Density (20°C) 0.836 to 0.844 g/ml
GC ≥95 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty