Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Patton-reeders reagent (Calconcarboxylic acid) indicator grade

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2,2'-Dihydroxy-4'-sulpho-1,1'-azonaphthalene-3-carboxylic acid3-Hydroxy-4-(2-hydroxy-4-sulpho-1-naphthylazo)naphthalene-2-carboxylic acid2-Hydroxy-1-(2-hydroxy-4-sulpho-1-naphthylazo)-3-naphthoic acid2-Hydroxy-1-(2-hydroxy-4-sulfo-1-naphthylazo)-3-naphthoic acidCal-Red®3-Hydroxy-4-(2-hydroxy-4-sulfo-1-naphthylazo)naphthalene-2-carboxylic acid2,2'-Dihydroxy-4'-sulfo-1,1'-azonaphthalene-3-carboxylic acid

15731-0100 15731-0500 15731-2500
TS15731-0500EA 162 USD InStock
TS15731-0500 TS15731-2500 TS15731-0100
Patton-reeders reagent (Calconcarboxylic acid) indicator grade
Patton-reeders reagent (Calconcarboxylic acid)


Formula: C₂₁H₁₄N₂O₇S
MW: 438.42 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00004078
CAS Number: 3737-95-9
EINECS: 223-117-0
Order Now
Specification Test Results
Appearance Dark purple to black Powder
Infrared spectrum Conforms
Suitability (for chel. det. of Ca) Passes test
Water ≤2.5 %
NMR (1H-NMR) Consistent with structure

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty