Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Silicone oil, 475 -525 mm²/s (25 °C) cP for melting point and boiling point apparatuses

Supplier: Thermo Scientific Chemicals
Synonyms: Polyphenyl-methylsiloxane

16385-0025 16385-0010 16385-5000
TS16385-5000EA 100.14 USD InStock
TS16385-5000 TS16385-0010 TS16385-0025
Silicone oil, 475 -525 mm²/s (25 °C) cP for melting point and boiling point apparatuses
Silicone oil


Melting Pt: −50 °C
Density: 0.96
Flash Pt: 300 °C
MDL Number: MFCD00132673
CAS Number: 68083-14-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty