Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroform 99+% stabilized for spectroscopy

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Methane, trichloro-TrichloroformTrichloromethane

Stabilization: Stabilised with amylene

16773-5000
TS16773-5000EA 69 USD InStock
TS16773-5000
Chloroform 99+% stabilized for spectroscopy
Chloroform


Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61 °C (760 mmHg)
Melting Pt: –63 °C
Density: 1.48 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
https://us-legacy.vwr.com/stibo/web/std.lang.all/69/36/726936.jpg
Order Now
Specification Test Results
Appearance Clear liquid
Water ≤0.05 %
GC ≥99.0 %
UV at 250 nm A: ≤0.5
UV at 260 nm A: ≤0.075
UV at 270 nm A: ≤0.01
UV at 280 nm A: ≤0.005
Color scale ≤10 APHA
Stabilizer 70 to 120 ppm Amylene
Residue after evaporation ≤0.0005 %
Acidity/alkalinity ≤0.0002 meq/g

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty