Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Dimethylacetamide 99+% for spectroscopy

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Acetic acid dimethylamideDimethyl Acetamide

16778-0010
TS16778-0010EA 169.54 USD InStock
TS16778-0010
N,N-Dimethylacetamide 99+% for spectroscopy
N,N-Dimethylacetamide


Formula: CH₃CON(CH₃)₂
MW: 87.12 g/mol
Boiling Pt: 164…166 °C (760 mmHg)
Melting Pt: –20 °C
Density: 0.937 g/cm³
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00008686
CAS Number: 127-19-5
EINECS: 204-826-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/13/2298913.jpg
Order Now
Specification Test Results
Appearance Clear liquid
Water ≤0.05 %
GC ≥99.0 %
UV at 270 nm A: ≤1
UV at 280 nm A: ≤0.3
UV at 290 nm A: ≤0.15
UV at 310 nm A: ≤0.05
UV at 320 nm A: ≤0.03
UV at 360 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty