Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

n-Heptane 95% for HPLC

Supplier: Thermo Scientific Chemicals
Danger

16782-0010
TS16782-0010EA 116 USD InStock
TS16782-0010
n-Heptane 95% for HPLC
n-Heptane


Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Boiling Pt: 98 °C (760 mmHg)
Melting Pt: –91 °C
Density: 0.683 g/cm³
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
UN: 1206
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/64/15/736415.jpg
Order Now
Specification Test Results
Appearance Clear liquid
Water ≤0.01 %
GC ≥94 %
UV at 200 nm A: ≤0.700
UV at 210 nm A: ≤0.301
UV at 227 nm A: ≤0.046
UV 1 cm cell vs HPLC-grade water
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %
Acidity ≤0.0002 meq/g
Filtered to 0.2 micrometer Filtered

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty