Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Silicone oil high temperature

Supplier: Thermo Scientific Chemicals
Synonyms: Polyphenyl-methylsiloxane

17466-5000 AC17466-1000 17466-0025
TS17466-5000EA 332.08 USD InStock
TS17466-5000 TS17466-0025 TS17466-1000
Silicone oil high temperature
Silicone oil


Density: 1.07 g/cm³
Flash Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00132673
CAS Number: 68083-14-7
Order Now
Specification Test Results
Appearance Clear colorless to straw yellow Oily liquid
Infrared spectrum Conforms
Refractive index 1.4940 to 1.5040 (20°C, 589 nm)
Viscosity 100 to 150 cSt (at 25°C)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty