Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium azide 99%, Extra Pure

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Sodium triazide

19038-1000 AC19038-0025 19038-5000 AC19038-0050
TS19038-0025EA 895.79 USD InStock
TS19038-0025 TS19038-0050 TS19038-1000 TS19038-5000
Sodium azide 99%, Extra Pure
Sodium azide


Formula: NaN₃
MW: 65.01 g/mol
Melting Pt: 275 °C
Density: 1.85 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003536
CAS Number: 26628-22-8
EINECS: 247-852-1
UN: 1687
ADR: 6.1,II
Merck Index: 12,08724
Order Now
Specification Test Results
Appearance White Crystalline powder
Assay ≥98.5 %
Loss on drying ≤0.5 %
pH ≥9 (5 % solution)
Sodium carbonate (Na2CO3) ≤0.15 %
Sodium hydroxide (NaOH) ≤0.1 %
Heavy metals (as Pb) ≤20 ppm
Insoluble matter ≤0.05 % (in water)

 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty