Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

cis-Platin 99.99% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: cis-Diaminodichloroplatinum(II)cis-Diamminedichloroplatinum (II)cis-Diamineplatinum(II) dichloridePlatinolcis-Dichlorodiammine platinum(II)Cisplatin

19376-0050 AC19376-2500 19376-0010
TS19376-2500EA 116 USD InStock
TS19376-2500 TS19376-0010 TS19376-0050
cis-Platin 99.99% (trace metals basis)
cis-Platin


Formula: H₆Cl₂N₂Pt
MW: 300.05 g/mol
Melting Pt: 277 °C
Storage Temperature: Ambient
MDL Number: MFCD00011623
CAS Number: 15663-27-1
EINECS: 239-733-8
UN: 3288
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Yellow to orange Powder or crystals
Infrared spectrum Conforms
Assay 64.0 to 66.0 % (Pt)
Total trace metal impurities ≤0.01 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty