Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphorus pentasulfide 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: di-Phosphorus pentasulphidedi-Phosphorus pentasulfide

AC19672-0010 AC19672-5000
TS19672-0010EA 88 USD InStock
TS19672-0010 TS19672-5000
Phosphorus pentasulfide 98+%
Phosphorus pentasulphide


Formula: P₂S₅
MW: 222.28 g/mol
Boiling Pt: 514 °C (760 mmHg)
Melting Pt: 282…286 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011441
CAS Number: 1314-80-3
EINECS: 215-242-4
UN: 1340
ADR: 4.3,II
Order Now
Specification Test Results
Appearance Yellow to green Powder
Assay ≥98.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty