Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eosin Y (yellowish), pure indicator grade

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Tetrabromofluorescein disodium saltEosin Y disodium saltEosin 2252′,4′,5′,7′-Tetrabromofluorescein disodium saltSolvent Red 43Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoateDisodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)Field's Stain B2,4,5,7-Tetrabromofluorescein disodium salt

20193-0250 20193-1000 20193-2500
TS20193-2500EA 172 USD InStock
TS20193-2500 TS20193-1000 TS20193-0250
Eosin Y (yellowish), pure indicator grade
Eosin Y (yellowish)


Formula: C₂₀H₆Br₄Na₂O₅
MW: 691.86 g/mol
Melting Pt: >300 °C (dec.)
Density: 1.018 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00005040
CAS Number: 17372-87-1
EINECS: 241-409-6
Order Now
Specification Test Results
Appearance Dark red-brown Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies
Loss on drying ≤10 %
UV Lambda max 511 to 519 nm
Specific extinction E (1%/1cm) ≥1100 (lambda max, 0.001M Na2CO3) on dried substance

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty