Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ascarite(II)® CO₂ Absorbent, 20 to 30 Mesh, Capacity: 40 to 50%, Product of Arthur H. Thomas Company, Thermo Scientific

Supplier: Thermo Scientific
ASCARITE®
20809-5000 20809-1000
TS20809-5000EA 665.3 USD InStock
TS20809-5000 TS20809-1000
Ascarite(II)® CO₂ Absorbent, 20 to 30 Mesh, Capacity: 40 to 50%, Product of Arthur H. Thomas Company, Thermo Scientific
Desiccators Desiccants
CAS No.: 81133-20-2

MDL No.: MFCD00132264
UN: 1823
Hazard Class: 8
Packing group: II
GHS Symbols: GHS05
Appearance: Beige to light brown Granules
Storage Temperature: Ambient
Signal word: Danger

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty