Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

beta-Alanyl-L-histidine

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

20817-0050 20817-0010 20817-1000 20817-0250
TS20817-0010EA 34 USD InStock
TS20817-0010 TS20817-1000 TS20817-0250 TS20817-0050
beta-Alanyl-L-histidine
Proteins and Peptides
  • Infrared spectrum: Conforms
  • Water: ≤1%
  • Specific optical rotation: 20.0 to 22.0° (20 °C, 589 nm) (c=2, water)
  • Melting point: 253 °C (dec.)

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty