Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Propionaldehyde 99+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Propanal

22051-1000 22051-5000 22051-0025
TS22051-5000EA 130.04 USD InStock
TS22051-5000 TS22051-0025 TS22051-1000
Propionaldehyde 99+%
Propionaldehyde


Formula: C₃H₆O
MW: 58.08 g/mol
Boiling Pt: 48 °C (760 mmHg)
Melting Pt: –81 °C
Density: 0.798 g/cm³
Flash Pt: –40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007020
CAS Number: 123-38-6
EINECS: 204-623-0
UN: 1275
ADR: 3,II
Merck Index: 12,08008
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3610 to 1.3660 (20°C, 589 nm)
GC ≥99.0 %
Color scale ≤20 APHA

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty