Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aspartame

Supplier: Thermo Scientific
22865-0050 22865-0250 22865-0010
TS22865-0050EA 185 USD InStock
TS22865-0050 TS22865-0250 TS22865-0010
Aspartame
Proteins and Peptides
  • Conforms: Infrared spectrum
  • Loss on drying: ≤4.5% (105 °C, 4 hrs)
  • UV transmittance: ≥95% (c=1, 2 N HCl, 430 nm) 1 cm cell
  • Specific optical rotation: +12.5 to +17.5° (20 °C, 589 nm) (c=4, 15 N formic acid) (within 30 min)
  • Heavy metals (as Pb): ≤10 ppm
  • Sulfated ash: ≤0.2%
  • Melting point: 248 to 250 °C

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty